Dimethyl sulphoxide-[D₆] ≥99.8% (Isotopic), MagniSolv™ for NMR spectroscopy, Sigma-Aldrich®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Dimethyl sulphoxide-[²H₆], Hexadeuterodimethyl sulphoxide, DMSO-D6, DMSO-d6, Dimethyl sulphoxide-d6

M-Clarity™ quality level = MQ100

1.03424.0009 1.03424.0010 1.03424.0100 1.03424.0025 1.03424.0005 1.03424.0011 1.03424.0050
1.03424.0009EA 60.5 EUR
1.03424.0009 1.03424.0010 1.03424.0100 1.03424.0025 1.03424.0005 1.03424.0011 1.03424.0050
Dimethyl sulphoxide-[D₆] ≥99.8% (Isotopic), MagniSolv™ for NMR spectroscopy, Sigma-Aldrich®
Dimethyl sulphoxide-[D₆]
Formule: D₃CS(O)CD₃
Kookpunt: 189 °C (5 mmHg)
Dichtheid: 1,19 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002090
CAS nummer: 2206-27-1
EINECS: 218-617-0

Order Now

Specification Test Results

Deuteration degree ≥ 99.80 %
Water (K. Fischer, H₂O + D₂O) ≤ 0.030 %
Performance test (NMR-spectrum) passes test
Water (NMR, H₂O) ≤ 0.020 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR