Cerium(IV) sulphate 0.05 mol/l (0.05 N) in aqueous solution, AVS TITRINORM volumetric solution

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Cerium disulphate, Ceric sulphate, Cerium(IV) sulphate

31307.294EA 198 EUR
31307.294
Cerium(IV) sulphate 0.05 mol/l (0.05 N) in aqueous solution, AVS TITRINORM volumetric solution
Cerium(IV) sulphate
Formule: Ce(SO₄)₂
MDL Nummer: MFCD00148852
CAS nummer: 13590-82-4
EINECS: 237-029-5
UN: 3264
ADR: 8,III
Merck Index: 13,02002

Order Now

Specification Test Results

Titer (20°C; real value 0.2 % accuracy) 0.0499 - 0.0501 mol/l
NIST traceable Confirmed


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR