VWR®, Buffertabletten voor hematologie
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
331552SEA 67.6 EUR
331552S 331562U 331932D 331942F
VWR®, Buffertabletten voor hematologie
Buffers Buffers voor microscopie
These tablets provide a rapid and convenient method of preparing a buffer solution.
Gurr buffer tablets for the preparation of microscopical stain solutions.
One tablet dissolved in distilled water, and made up to 100 ml, produces a solution of the stated pH at 20 °C. The molarity of the buffer solutions is 0,005 M.