Chromium standard solution, 10000 mg/L Cr in 5% nitric acid (from Cr(NO₃)₃), standard for ICP, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
38722.AE 38722.AD 38722.AP
38722.AEEA 161 EUR
38722.AE 38722.AD 38722.AP
Chromium standard solution, 10000 mg/L Cr in 5% nitric acid (from Cr(NO₃)₃), standard for ICP, Specpure®
Standaarden Kationische standaarden Chroom standaarden
Molecular Formula: Cr(NO₃)₃ in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00010944
Certificaten: Traceable to NIST