pH/ion meter, SevenDirect SD50
Leverancier: Mettler - Toledo
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
30671544 30671545 30671546 30671547 30671548 30671549 30671552
662-2384EA 1720 EUR
662-2384 662-2385 662-2386 662-2387 662-2388 662-2389 662-2392
pH/ion meter, SevenDirect SD50
Elektrochemische meters pH- en multiparametersystemen
Robust bench top meter for accurate pH and ion determinations in the lab and near production.
  • Large colour touch screen provides direct measurement and easy data entry
  • Instructions guide users through the sensor calibration process
  • Stores data, such as time stamp, sample and sensor ID with every measurement and prints or exports records automatically
  • Closed connector compartment and replaceable cover keep the instrument safe from dust and spills, even with sensors attached (IP 54 in-use)
The large colour touch screen offers direct menu access, fastMore Product Information