Nitrilotriacetic acid 99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: NTA, 2,2',2''-Nitrilotriacetic acid

415740010 415740050 415742500 415740051
ACRO415740010EA 224 EUR
ACRO415740010 ACRO415740050 ACRO415742500 ACRO415740051
Nitrilotriacetic acid 99%
Nitrilotriacetic acid
Formule: C₆H₉NO₆
Kookpunt: 498 °C (1013 hPa)
Dichtheid: 1,67 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004287
CAS nummer: 139-13-9
EINECS: 205-355-7
ADR: 8,II
Merck Index: 12,06675

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Titration excess NaOH ≥98.5 %
Titration with NaOH ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR