Neopentyl bromide 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: 2,2-Dimethyl-1-propyl bromide, 1-Bromo-2,2-dimethylpropane

425720250 425720010 425720050
ACRO425720250EA 190 EUR
ACRO425720250 ACRO425720010 ACRO425720050
Neopentyl bromide 98%
Neopentyl bromide
Formule: C₅H₁₁Br
Kookpunt: 105…106 °C (767 mmHg)
Dichtheid: 1,199 g/cm³
Vlampunt: 7 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00000209
CAS nummer: 630-17-1
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4350 to 1.4390 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR