2,4-Dimethoxy-1-fluorobenzene 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC450147-10G PC450147-1G PC450147-5G PC450147-25G
APOSPC450147-10GEA 0 EUR
APOSPC450147-10G APOSPC450147-1G APOSPC450147-5G APOSPC450147-25G
2,4-Dimethoxy-1-fluorobenzene 97%
2,4-Dimethoxy-1-fluorobenzene
Formule: C₈H₉FO₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00042265
CAS nummer: 17715-70-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR