Laboratory Chairs with Castors, Nexxit 2
Leverancier: bimos
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
9033-MG01-3285 9033-SP01-3280 9033-MG01-3277 9033-SP01-3277 9033-2000-3277 9033-2000-3285 9033MG01521ESD
BIMO9033-MG01-3285EA 387 EUR
BIMO9033-MG01-3285 BIMO9033-SP01-3280 BIMO9033-MG01-3277 BIMO9033-SP01-3277 BIMO9033-2000-3277 BIMO9033-2000-3285 BIMO9033MG01521ESD
Laboratory Chairs with Castors, Nexxit 2
Stoelen
Nexxit is the perfect fit for all applications in industry, for ESD workstations and for the laboratory.
  • Synchronous mechanism with automatic weight regulation
  • Seat height 450 to 600 mm
  • Ergonomic package composed of:
  • Seat depth adjustment, adjustable seat inclination
  • and backrest adjustable in height
  • Steel base
  • Useful functional handles
  • Support for a strong back
  • Work with added comfort
  • Automatically well seated
  • Relieves strain on the arms
  • Robust
Ergonomic support, practical handling and outstanding robustness - modern industry demands specificMore Product Information