Anti-SLC25A10 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Leverancier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS-3997R-A555
BOSSBS-3997R-A555EA 893 EUR
BOSSBS-3997R-A555
Anti-SLC25A10 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antilichamen
Involved in translocation of malonate, malate and succinate in exchange for phosphate, sulfate, sulfite or thiosulfate across mitochondrial inner membrane.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SLC25A10
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR