Anti-p-Cdk1/Cdc2 Rabbit Polyclonal Antibody [clone: T14 ]

Leverancier: Bioworld Technology
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS6341
BWRLBS6341EA 379 EUR
BWRLBS6341
Anti-p-Cdk1/Cdc2 Rabbit Polyclonal Antibody [clone: T14 ]
Antilichamen
Synthetic phosphopeptide derived from human Cdk1/Cdc2 around the phosphorylation site of Serine 795.

p-Cdk1/Cdc2 (T14 ) pAb detects endogenous levels of Cdk1/Cdc2 protein only when phosphorylated at Thr14.

Type: Primary
Antigen: p-Cdk1/Cdc2
Clonality: Polyclonal
Clone: T14
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR