Human recombinant Shp-2 (from E. coli)
Leverancier: ENZO LIFE SCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BMLSE2730020
ENZOBMLSE2730020EA 548 EUR
ENZOBMLSE2730020
Human recombinant Shp-2 (from E. coli)
Eiwitten en peptiden
Produced in E. coli. full length human Shp-2 (aa 1-593).