Anti-IL7R Mouse Monoclonal Antibody (Alexa Fluor® 546) [clone: R34-34]

Leverancier: ENZO LIFE SCIENCES
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

ENZABS2500100
ENZOENZABS2500100EA 653 EUR
ENZOENZABS2500100
Anti-IL7R Mouse Monoclonal Antibody (Alexa Fluor® 546) [clone: R34-34]
Antilichamen
Recommended Applications: IF

Species reactivity: Human, Dog

Type: Primary
Antigen: IL7R
Clonality: Monoclonal
Clone: R34-34
Conjugation: Alexa Fluor® 546
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR