Leucosep™ tubes with porous barrier and screw cap
Leverancier: Greiner Bio-One
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
227290_300 163289_500 227289_300 163290_500 163288_500 227288_250
720-1840EA 945 EUR
720-1840 GREI163289_500 GREI227289_300 GREI163290_500 GREI163288_500 GREI227288_250
Leucosep™ tubes with porous barrier and screw cap
Diagnostische systemen Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
Leucosep™ tubes have been developed for the optimal separation of lymphocytes and peripheral mononuclear cells from human whole blood and bone marrow by means of density gradient centrifugation.
  • Simplified filling through transparent porous barrier of high-grade polyethylene
  • Rapid separation in 15 minutes at room temperature
  • Removal of erythrocytes and granulocytes, no recontamination with erythrocytes, no blocking of marker molecules
  • Sterile tubes
  • Tubes conical bottom, graduated, made of polypropylene
  • Designed for single-use only
The key feature ofMore Product Information