MES sodium salt, white powder

Leverancier: MP Biomedicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: MES-Na

0215245410
ICNA0215245410EA 32.5 EUR
ICNA0215245410
MES sodium salt, white powder
MES sodium salt

Solubility (10% aq soln): clear solution.


Solubility (10% aq soln): clear solution.


Opgelet: Research Use Only (RUO).

Formule: C₆H₁₂NNaO₄S
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00065473
CAS nummer: 71119-23-8

Order Now

Specification Test Results

Identity Test Passes
FTIR Conforms to Standard
pH (1% aq. soln) 8.5 - 10.4
Moisture Content ≤9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR