Tubing for chromatography, ETFE
Leverancier: VICI JOUR RESEARCH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
JR-T-083-M3 JR-T-084-M3 JR-T-085-M3 JR-T-086-M3 JR-T-087-M3 JR-T-088-M3 JR-T-084-M10 JR-T-086-M25 JR-T-086-M10 JR-T-085-M25 JR-T-085-M10 JR-T-084-M25 JR-T-083-M25 JR-T-083-M10 JR-T-082-M25 JR-T-082-M10 JR-T-080-M25 JR-T-080-M10 JR-T-087-M1 JR-T-088-M1
554-1022EA 70.1 EUR
554-1022 JOURJR-T-084-M3 JOURJR-T-085-M3 JOURJR-T-086-M3 JOURJR-T-087-M3 JOURJR-T-088-M3 JOURJR-T-084-M10 JOURJR-T-086-M25 JOURJR-T-086-M10 JOURJR-T-085-M25 JOURJR-T-085-M10 JOURJR-T-084-M25 JOURJR-T-083-M25 JOURJR-T-083-M10 JOURJR-T-082-M25 JOURJR-T-082-M10 JOURJR-T-080-M25 JOURJR-T-080-M10 JOURJR-T-087-M1 JOURJR-T-088-M1
Tubing for chromatography, ETFE
Slangen Chromatography Tubing
ETFE.
  • Ideal for applications with chloride salts, which typically corrode stainless steel
  • Higher burst pressure than PTFE
Tubing has some outstanding properties which make it particulary suitable for HPLC.