VWR® Buffer, MOPS-SDS, 20X, UltraPure Grade
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
K855-500MLEA 120 EUR
K855-500ML
VWR® Buffer, MOPS-SDS, 20X, UltraPure Grade
Buffers Buffers voor elektroforese Running-buffers
Useful for high resolution of proteins on neutral pH SDS-PAGE gels.
  • Ultra-pure grade
  • Convenient concentrations and packaging
  • Good quality, consistency and resolution