Portable photometers, MultiDirect
Leverancier: Lovibond
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
210000
LOVI210000EA 1571.4 EUR
LOVI210000
Portable photometers, MultiDirect
Colorimeters Colorimeters met meerdere Parameters
The MultiDirect is a microprocessor controlled photometer with ergonomically designed keypad and large format graphic display. It is equipped with preprogrammed methods based on Lovibond® tablet, liquid and powder reagents.
  • High accuracy from dual beam technology
  • Instrument and sample chamber is waterproof
  • Long term stable LED light sources
  • Update of new methods and languages via the internet
Leveringsinformatie: Supplied with rechargeable batteries, mains charger, PC connection cable, carrying case, 3× round vials 24 mm, 3× round vials 16 More Product Information