Anti-CUL-3 Rabbit polyclonal antibody

Leverancier: Novus Biologicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

NB100-58788
NOVUNB100-58788EA 684 EUR
NOVUNB100-58788
Anti-CUL-3 Rabbit polyclonal antibody
Antilichamen
The Cullin 3 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Cullin 3. This antibody reacts with human, mouse. The Cullin 3 Antibody has been validated for the following applications: Western Blot, Immunoprecipitation, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: CUL-3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR