Anti-PECAM1 Mouse Monoclonal Antibody [clone: 3G4]

Leverancier: OriGene
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

CF504811
ORIGCF504811EA 833 EUR
ORIGCF504811
Anti-PECAM1 Mouse Monoclonal Antibody [clone: 3G4]
Antilichamen
Recommended Dilutions: Western Blot: 1:200-1:1000; Immunochemistry: 1:150

Type: Primary
Antigen: PECAM1
Clonality: Monoclonal
Clone: 3G4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR