Density hydrometers
Leverancier: Ludwig Schneider GmbH & CO. KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
3031001 3031002 3031003 3031004 3031005 3031006 3031007 3031008 3031009 3031010 3031011 3031012 3031013 3031014
SCNL3031001EA 20.3 EUR
SCNL3031001 SCNL3031002 SCNL3031003 SCNL3031004 SCNL3031005 SCNL3031006 SCNL3031007 SCNL3031008 SCNL3031009 SCNL3031010 SCNL3031011 SCNL3031012 SCNL3031013 SCNL3031014
Density hydrometers
Hydrometers
Density hydrometers calibrated at 20 °C.
* Works certificate.
Bestelinformatie: Available with works or DAkkS calibration certificate on request.
Verpakking: Packed in a square plastic sleeve.