Bottles, narrow neck, round, PFA, with screw caps
Leverancier: SEMADENI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
3840
SEMA3840EA 256 EUR
SEMA3840
Bottles, narrow neck, round, PFA, with screw caps
Flessen Kunststof flessen
PFA, translucent.
  • Very good chemical resistance
  • Very good temperature resistance