Dicyclohexyl sulphide ≥90.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

D1617-25ML D1617-500ML
TCIAD1617-25MLEA 26.4 EUR
TCIAD1617-25ML TCIAD1617-500ML
Dicyclohexyl sulphide ≥90.0% (by GC)
Dicyclohexyl sulphide
Formule: C₁₂H₂₂S₂
Kookpunt: 110°C
Dichtheid: 1,0457 g/cm³ (20 °C)
MDL Nummer: MFCD00013759
CAS nummer: 2550-40-5
EINECS: 219-851-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d4f90b16c8b456ed6b4aeebdb3790e66d4a8ac2b&vid=d4f90b16c8b456ed6b4aeebdb3790e66d4a8ac2b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 90.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR