L(-)-Tyrosine-[¹⁵N]

Leverancier: BACHEM BIOCHEMICA
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

4026141.0100 4026141.0250 4026141.1000
BACE4026141.0100EA 0 EUR
BACE4026141.0100 BACE4026141.0250 BACE4026141.1000
L(-)-Tyrosine-[¹⁵N]
L(-)-Tyrosine-[¹⁵N]
Formule: C₉H₁₁NO₃
MDL Nummer: MFCD00055773
CAS nummer: 35424-81-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR