3-Pentadecylphenol ≥90.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
P1202-25G
P1202-100G
TCIAP1202-25GEA
28.6
EUR
TCIAP1202-25G
TCIAP1202-100G
3-Pentadecylphenol ≥90.0% (by GC)
3-Pentadecylphenol
Formule:
C₂₁H₃₆O Kookpunt: 232°C |
MDL Nummer:
MFCD00002310 CAS nummer: 501-24-6 EINECS: 207-921-9 |
|
Specification Test Results
Appearance | White to Gray to Brown powder to crystal |
Purity (GC) | min. 90.0 % |
Melting point | 49.0 to 53.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...