Adapters
Leverancier: VICI JOUR RESEARCH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
JR-0611 JR-0612 JR-0617 JR-0622 JR-0623 JR-5505 JR-5595 JR-0625 JR-0626 JR-062
546-1251EA 50.9 EUR
546-1251 554-1003 554-1008 554-1013 554-1014 JOURJR-5505 546-1383 554-1016 554-1017 JOURJR-062
Adapters
Fittingen en connectoren
These PEEK adapters connect standard LC fittings to standard LC ports, usable with most common organic and inorganic HPLC solvents without risking corrosion, metal contamination, or protein degradation.
  • Easy connection of solvent lines
  • Low dead volume
  • Completely inert and biocompatible
Pressure rating of M6, 1/4–28 is <70 bar (1000 psi), Luer is <35 bar (509 psi) and varies with tubing material and ID.