Mono slot samplers, SteriWare®
Leverancier: Sampling Systems
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
8069H-1000 8069H-500 8069H-1000S 8069H-500S
SAMP8069H-1000EA 321 EUR
SAMP8069H-1000 SAMP8069H-500 SAMP8069H-1000S SAMP8069H-500S
Mono slot samplers, SteriWare®
Monsternemers Monsternemers voor eenmalig gebruik
Disposable samplers for taking a single sample from free flowing powder and granules.
  • Assembled and packed in a class 100 000 cleanroom
  • BSE/TSE free
  • Full batch traceability
Designed for pharmaceutical and other hygiene conscious industries.
Certificaten: FDA, EU 10/2011 and EC 1935/2004. Approved for use in ATEX zones.
Verpakking: Individually bagged.