Polyethylene glycol 600
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
Ethylene glycol polymer average Mn 600, PEG 600
B21798.0B
B21798.30
B21798.0BEA
71.9
EUR
B21798.0B
B21798.30
Polyethylene glycol 600
Polyethylene glycol 600
CAS No.: 25322-68-3
Molecular Formula: H(OCH2CH2)nOH
Flash Point: 252°(485°F)
Physical Form: Low-melting solid
Density: 1.120
Refractive Index: 1.4680
Odour: Odourless
pH: 5-7
Melting Point: 17-23°
Merck Reference: 14,7568
MDL No.: MFCD00081839
Formule:
H(OCH₂CH₂)nOH Kookpunt: ∼250 °C (1013 hPa) Smeltpunt: 17…23 °C Dichtheid: 1,120 Vlampunt: 252 °C (485 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00081839 CAS nummer: 25322-68-3 EINECS: 500-038-2 Merck Index: 14,7568 |