di-Ammonium oxalate monohydrate, Supelco®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Ammonium oxalate monohydrate, Ammonium oxalate-1-hydrate, Oxalic acid diammonium salt monohydrate, Oxalic acid ammonium salt monohydrate, Ammonium ethanedioate monohydrate, Ammonium Oxalate, Monohydrate
M-Clarity™ quality level = MQ200
1.01190.1000 1.01190.9050
1.01190.1000EA 146 EUR
1.01190.1000 1.01190.9050
di-Ammonium oxalate monohydrate, Supelco®
di-Ammonium oxalate monohydrate
Formule: (NH₄)₂C₂O₄·H₂O
Kookpunt: 70 °C (1013 hPa)
Smeltpunt: 70 °C
Dichtheid: ∼1.5 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149694
CAS nummer: 6009-70-7
EINECS: 214-202-3
UN: 3288
ADR: 6.1,III
Merck Index: 13,00541