tetra-Chloroauric(III) acid trihydrate

Leverancier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Synoniemen: Gold(III) chloride trihydrate, Gold Chloride hydrate, Hydrogen tetrachloroaurate(III) trihydrate, Hydrogen tetrachloroaurate (III) trihydrate, tetra-Chloroauric acid trihydrate

3867.1 3867.2
ROTH3867.1EA 0 EUR
ROTH3867.1 ROTH3867.2
tetra-Chloroauric(III) acid trihydrate
tetra-Chloroauric(III) acid trihydrate
Formule: HAuCl₄·3H₂O
Smeltpunt: 30 °C
Dichtheid: 3,9 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149904
CAS nummer: 16961-25-4
EINECS: 240-948-4
UN: 3260
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR