Tributyl citrate 99+%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Citric acid tributyl ester, Hydroxytricarballylic acid tributyl ester, Tributyl 2-hydroxypropane-1,2,3-tricarboxylate
421440010 421442500
ACRO421440010EA 179 EUR
ACRO421440010 ACRO421442500
Tributyl citrate 99+%
Tributyl citrate
Formule: C₁₈H₃₂O₇
Kookpunt: 325 °C (760 mmHg)
Smeltpunt: –20 °C
Dichtheid: 1,042 g/cm³
Vlampunt: 157 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00027217
CAS nummer: 77-94-1
EINECS: 201-071-2
Merck Index: 13,01563
Order Now
Specification Test Results
Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4440 to 1.4460 (20°C, 589 nm)
Water ≤0.3 % (K.F.)
GC ≥99.0 %
Color scale ≤50 APHA
Acidity ≤0.2 mg KOH/g