Aluminium ammonium sulphate dodecahydrate

Leverancier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: Aluminium ammonium bis(sulphate) dodecahydrate, Ammonium alum, Ammonium aluminum sulphate dodecahydrate, Aluminum ammonium sulphatedodecahydrate

X862.1 X862.2
ROTHX862.1EA 41.5 EUR
ROTHX862.1 ROTHX862.2
Aluminium ammonium sulphate dodecahydrate
Aluminium ammonium sulphate dodecahydrate
Formule: AlNH₄(SO₄)₂·12H₂O
Kookpunt: 200 °C (1013 hPa)
Smeltpunt: 93 °C
Dichtheid: 1,64 g/cm³ (20 °C)
MDL Nummer: MFCD00051042
CAS nummer: 7784-26-1
EINECS: 232-055-3
Merck Index: 13,00325

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR