4-Benzylamino-1-butanol ≥96.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

B1599-25G B1599-5G
TCIAB1599-25GEA 96.8 EUR
TCIAB1599-25G TCIAB1599-5G
4-Benzylamino-1-butanol ≥96.0% (by GC)
4-Benzylamino-1-butanol
Formule: C₁₁H₁₇NO
Kookpunt: 140°C
MDL Nummer: MFCD00191609
CAS nummer: 59578-63-1

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR