3-Bromobenzhydrazide ≥98.0% (by GC, titration analysis)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
B4148-1G B4148-5G
TCIAB4148-1GEA 22 EUR
TCIAB4148-1G TCIAB4148-5G
3-Bromobenzhydrazide ≥98.0% (by GC, titration analysis)
3-Bromobenzhydrazide
Formule: C₇H₇BrN₂O
MDL Nummer: MFCD00007600
CAS nummer: 39115-96-3
EINECS: 254-298-4
Order Now
Specification Test Results
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 155.0 to 159.0 deg-C