2-Fluoro-5-iodobenzotrifluoride ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: 1-Fluoro-4-iodo-2-(trifluoromethyl)benzene
F1161-25G F1161-5G
TCIAF1161-25GEA 130 EUR
TCIAF1161-25G TCIAF1161-5G
2-Fluoro-5-iodobenzotrifluoride ≥98.0% (by GC)
2-Fluoro-5-iodobenzotrifluoride
Formule: C₇H₃F₄I
MDL Nummer: MFCD00236678
CAS nummer: 59382-39-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b8271a5c3455dbab600f3f77b391f36fcc09f4ba&vid=b8271a5c3455dbab600f3f77b391f36fcc09f4ba&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity (GC) min. 98.0 %