2-Phenylpropionitrile ≥90.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
P0860-1ML P0860-10ML
TCIAP0860-1MLEA 19.8 EUR
TCIAP0860-1ML TCIAP0860-10ML
2-Phenylpropionitrile ≥90.0% (by GC)
2-Phenylpropionitrile
Formule: C₉H₉N
Kookpunt: 230°C
Vlampunt: 94°C
MDL Nummer: MFCD00001871
CAS nummer: 1823-91-2
EINECS: 217-354-9
UN: 3276
ADR: 6.1,III
Order Now
Specification Test Results
Purity (GC) min. 90.0 %