Y5, Organic Photovoltaics (OPV) reagent

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

H66460.MC H66460.MD H66460.ME
H66460.MCEA 673 EUR
H66460.MC H66460.MD H66460.ME
Y5, Organic Photovoltaics (OPV) reagent
Y5
Formule: C₈₂H₉₀N₈O₂S₅
CAS nummer: 2304444-48-0

Order Now

Specification Test Results

Appearance (Color) Black
Form Flake or threadlike
Bandgap energy 1,41 ± 0,02 eV


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR