tert-Butyl 4-bromo-1H-indazole-5-carboxylate

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

OR305101-250MG OR305101-1G
APOSOR305101-250MGEA 67.7 EUR
APOSOR305101-250MG APOSOR305101-1G
tert-Butyl 4-bromo-1H-indazole-5-carboxylate
tert-Butyl 4-bromo-1H-indazole-5-carboxylate
Formule: C₁₂H₁₃BrN₂O₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD22572308
CAS nummer: 1203662-37-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR