6-Fluoro-2-methyl-2,4-dihydro-1,4-benzoxazin-3-one 98%
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PC910918-1G
PC910918-5G
APOSPC910918-1GEA
330
EUR
APOSPC910918-1G
APOSPC910918-5G
6-Fluoro-2-methyl-2,4-dihydro-1,4-benzoxazin-3-one 98%
6-Fluoro-2-methyl-2,4-dihydro-1,4-benzoxazin-3-one
Formule:
C₉H₈FNO₂ Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD08446952 CAS nummer: 925005-78-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...