Benzo[d]isoxazol-3-ol

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 3-Hydroxybenzisoxazole, 3-Hydroxy-1,2-benzisoxazole

OR27158-10G OR27158-1G OR27158-5G
APOSOR27158-10GEA 220 EUR
APOSOR27158-10G APOSOR27158-1G APOSOR27158-5G
Benzo[d]isoxazol-3-ol
Benzo[d]isoxazol-3-ol
Formule: C₇H₅NO₂
Smeltpunt: 137…140 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00125030
CAS nummer: 21725-69-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR