ZHE Hazardous Waste System
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
YY1301015 YY2209064 YY2209068 YT3009001 YT3009002 YT3009003 YT3009013 YT3009014 YT3009015 YT3009016 YT3009017 YT3009018 YT30090HW
MILFYY1301015EA 323 EUR
MILFYY1301015 MILFYY2209064 MILFYY2209068 MILFYT3009001 MILFYT3009002 MILFYT3009003 MILFYT3009013 MILFYT3009014 MILFYT3009015 MILFYT3009016 MILFYT3009017 MILFYT3009018 MILFYT30090HW
ZHE Hazardous Waste System
Filters Membraanfilters
ZHE (Zero Head Space Extractor) hazardous waste filtration system, which is approved by the US Environmental Protection Agency (EPA) as an apparatus for Toxicity Characteristic Leaching Procedure (TCLP) measurement. The system cannot pressurise unless completely assembled; this eliminates accidentally 'shooting the piston' out of the unit and assures safe operation. Choose both manual and automatic venting modes.
  • Allows for pressursation of filter without introducing air, due to movable piston design
  • Prevents loss of volatiles
  • Manual and automatic venting modes
  • System cannot be pressurised unless completely assembled, thereby preventing 'shooting the piston' out of the unit accidentally