Celtellers, Sedgewick Rafter
Leverancier: MARIENFELD
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
0611300 0611400
MARI0611300EA 811 EUR
MARI0611300 MARI0611400
Celtellers, Sedgewick Rafter
Object- en dekglaasjes Telkamers
Counting chambers according to Sedgewick for particles and microorganisms in transparent liquids.
A cell is ruled in a grid subdividing 1,0 ml into 1000 μl.
Leveringsinformatie: Supplied with a cover glass.