Bariumchloride, anhydrous ≥99.998% (metals basis)
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
10995.02
10995.06
10995.22
10995.02EA
44.3
EUR
10995.02
10995.06
10995.22
Bariumchloride, anhydrous ≥99.998% (metals basis)
Bariumchloride
Purity: 99.998%
CAS No.: 10361-37-2
Molecular Formula: BaCl2
Formula Weight: 208.25
Physical Form: Powder, may contain small lumps
Density: 3.9
Appearance: White
Odour: Odourless
Melting Point: 963°
Boiling Point: 1560°
Merck Reference: 14,971
UN No.: 1564
MDL No.: MFCD00003445
Formule:
BaCl₂ Kookpunt: 1560 °C Smeltpunt: 963 °C Dichtheid: 3,9 Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00003445 CAS nummer: 10361-37-2 EINECS: 233-788-1 UN: 1564 ADR: 6.1,III Merck Index: 14,971 |