Ethyl-3,3-diethoxypropionate 98%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
269640050 269640250 269641000
ACRO269640050EA 24.2 EUR
ACRO269640050 ACRO269640250 ACRO269641000
Ethyl-3,3-diethoxypropionate 98%
Ethyl-3,3-diethoxypropionate
Formule: C₉H₁₈O₄
Kookpunt: 95 °C (17 mmHg)
Dichtheid: 0,97 g/cm³
Vlampunt: 93 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00009865
CAS nummer: 10601-80-6
EINECS: 234-223-1
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4100 to 1.4120 (20°C, 589 nm)
GC ≥97.5 %