Ethyl trichloroacetate ≥97.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
T1504-25G
T1504-500G
TCIAT1504-25GEA
23.1
EUR
TCIAT1504-25G
TCIAT1504-500G
Ethyl trichloroacetate ≥97.0% (by GC)
Ethyl trichloroacetate
Formule:
C₄H₅Cl₃O₂ Kookpunt: 167°C Dichtheid: 1,38 g/cm³ (20 °C) Vlampunt: 65°C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00000795 CAS nummer: 515-84-4 EINECS: 208-212-7 ADR: 6.1,I Merck Index: 14,09627 |
|
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...