Anti-IPPK/IPK1 Rabbit Monoclonal Antibody [clone: EPR20893-54]
Leverancier: Abcam
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AB230807-100 AB230807-1ML
ABCAAB230807-100EA 763 EUR
ABCAAB230807-100 ABCAAB230807-1ML
Anti-IPPK/IPK1 Rabbit Monoclonal Antibody [clone: EPR20893-54]
Antilichamen
Rabbit monoclonal [EPR20893-54] to IPPK/IPK1.
Opgelet: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: IPPK/IPK1
Clonality: Monoclonal
Clone: EPR20893-54
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human