The Avantor XT96 App
The Avantor XT96 app is a mobile solution for controlling XT96 and XTender96 thermal cyclers.
The app enables you to write new PCR protocols, start, stop and check on your PCR run
(remaining time, current temperature cycle, etc.) via remote.
The app is also designed for multi-control of several thermal cyclers: just connect to all the
cyclers in your lab and control them all with one mobile device.