Aluminium ≥99.9995% (metals basis), foil 50x125 mm (2.0x4.9 in), Thickness: 0.13 mm (0.005 in)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Aluminum

00004.KF 00004.KI
00004.KFEA 81.9 EUR
00004.KF 00004.KI
Aluminium ≥99.9995% (metals basis), foil 50x125 mm (2.0x4.9 in), Thickness: 0.13 mm (0.005 in)
Aluminium

Purity: 99.9995% CAS No.: 7429-90-5 Physical Form: ≈2.2g/50x125mm Odour: Odourless MDL No.: MFCD00134029

Formule: Al
MW: 26.98 g/mol
Kookpunt: 2327
Smeltpunt: 660
Dichtheid: 2.7 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00134029
CAS nummer: 7429-90-5
EINECS: 231-072-3
Merck Index: 13,00321

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR