Copper ≥99.999% (metals basis), draad, Puratronic® oxygen-free, Ø 0.64 mm (0.025 in)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

00098.G2 00098.G6
00098.G2EA 74.7 EUR
00098.G2 00098.G6
Copper ≥99.999% (metals basis), draad, Puratronic® oxygen-free, Ø 0.64 mm (0.025 in)
Copper

Purity: 99.999% CAS No.: 7440-50-8 Physical Form: ≈2.87g/m Odour: Odourless MDL No.: MFCD00010965

Formule: Cu
Kookpunt: 2595 °C (1013 hPa)
Smeltpunt: 1083 °C
Dichtheid: 8,94 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00010965
CAS nummer: 7440-50-8
EINECS: 231-159-6
Merck Index: 13,02545

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR