Gold ≥99.99% (metals basis), foil, Premion®, Thickness: 0.127 mm (0.005 in)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

00132.FF 00132.FI
00132.FFEA 477 EUR
00132.FF 00132.FI
Gold ≥99.99% (metals basis), foil, Premion®, Thickness: 0.127 mm (0.005 in)
Gold

Purity: 99.99% CAS No.: 7440-57-5 Physical Form: ≈1.5g/25x25mm Odour: Odourless MDL No.: MFCD00003436

Formule: Au
Kookpunt: 2800 °C (1013 hPa)
Smeltpunt: 1063 °C
Dichtheid: 19,3 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003436
CAS nummer: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR